Anti-GCNT3 Rabbit Polyclonal Antibody

Supplier: Prosci

71-082
10080-584EA 1042.19 CAD
10080-584
Anti-GCNT3 Rabbit Polyclonal Antibody
Antibodies
GCNT3 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 5 - 7.5 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GCNT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Dog, Gorilla, Horse, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR