Anti-CADM2 Rabbit Polyclonal Antibody

Supplier: Prosci
71-087
10080-594EA 1078.35 CAD
10080-594
Anti-CADM2 Rabbit Polyclonal Antibody
Antibodies
CADM2 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 - 15 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CADM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Gibbon, Bovine, Dog, Gorilla, Horse, Human, Monkey, Pig, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR