Anti-STEAP2 Rabbit Polyclonal Antibody

Supplier: Prosci

71-101
10080-622EA 983.75 CAD
10080-622
Anti-STEAP2 Rabbit Polyclonal Antibody
Antibodies
STEAP2 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: STEAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Gibbon, Chimpanzee, Dog, Guinea pig, Gorilla, Hamster, Horse, Human, Monkey, Mouse, Orangutan, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR