Anti-SLC1A4 Rabbit Polyclonal Antibody

Supplier: Prosci

71-121
10077-582EA 1042.19 CAD
10077-582
Anti-SLC1A4 Rabbit Polyclonal Antibody
Antibodies
SLC1A4 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 - 12.5 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SLC1A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Dog, Gorilla, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR