Order Entry
ContactUsLinkComponent
Anti-ERAP1 Rabbit Polyclonal Antibody
Anti-ERAP1 Rabbit Polyclonal Antibody
Catalog #: 10077-586
Supplier:  Avantor
Anti-ERAP1 Rabbit Polyclonal Antibody
Catalog #: 10077-586
Supplier:  Avantor
Discontinued
Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    endoplasmic reticulum aminopeptidase 1
  • Clonality:
    Polyclonal
  • Gene ID:
    Q9NZ08
  • Host:
    Rabbit
  • Reactivity:
    Monkey,
    Gibbon,
    Gorilla,
    Human
  • Antigen symbol:
    ERAP1
  • Conjugation:
    Unconjugated
  • ImmunoChemistry:
    Yes
  • Size:
    0.05 mg
  • Antigen synonyms:
    ERAP1
  • Storage buffer:
    PBS, 0.1% sodium azide.
  • Storage temperature:
    Antibody may be stored short term at 4 °C, long term aliquot and store at –20 °C, avoid freeze thaw cycles.
  • Concentration:
    1 mg/mL
  • Shipping temperature:
    4 °C
  • Immunogen:
    Synthetic 15 amino acid peptide from internal region of human ERAP1 / ARTS1.
  • Tested applications:
    Immunoaffinity Chromatography
  • Purification:
    Immunoaffinity Chromatography
  • Cat. no.:
    10077-586

Specifications

About this item

ERAP1 / ARTS1 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 - 12.5 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ERAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human, Monkey

Customers Who Bought This Also Bought