Anti-GM2A Rabbit Polyclonal Antibody

Supplier: Prosci

71-128
10077-596EA 983.75 CAD
10077-596
Anti-GM2A Rabbit Polyclonal Antibody
Antibodies
GM2A Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 5 - 7.5 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GM2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR