Anti-C14ORF1 Rabbit Polyclonal Antibody

71-136
10077-612EA 1042.19 CAD
10077-612
Anti-C14ORF1 Rabbit Polyclonal Antibody
Antibodies
C14orf1 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 - 15 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: C14orf1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Chimpanzee, Dog, Guinea pig, Gorilla, Hamster, Horse, Human, Monkey, Mouse, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR