Anti-ITGA6 Rabbit Polyclonal Antibody

Supplier: Prosci
71-151
10080-682EA 983.75 CAD
10080-682
Anti-ITGA6 Rabbit Polyclonal Antibody
Antibodies
ITGA6 / CD49f Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 1 - 5 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ITGA6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Gibbon, Gorilla, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR