Anti-SLC5A6 Rabbit Polyclonal Antibody

Supplier: Prosci

71-157
10080-694EA 983.75 CAD
10080-694
Anti-SLC5A6 Rabbit Polyclonal Antibody
Antibodies
  • SLC5A6 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 - 15 mg/ml.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SLC5A6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR