Anti-WNT5B Rabbit Polyclonal Antibody

71-158
10080-696EA 1078.68 CAD
10080-696
Anti-WNT5B Rabbit Polyclonal Antibody
Antibodies
WNT5B Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: WNT5B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Dog, Gorilla, Horse, Human, Orangutan, Pig, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR