Anti-ERP44 Rabbit Polyclonal Antibody

Supplier: Prosci

71-185
10080-750EA 1042.19 CAD
10080-750
Anti-ERP44 Rabbit Polyclonal Antibody
Antibodies
ERP44 / TXNDC4 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 - 15 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ERP44
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR