Anti-GLG1 Rabbit Polyclonal Antibody

Supplier: Prosci

71-204
10080-788EA 1078.68 CAD
10080-788
Anti-GLG1 Rabbit Polyclonal Antibody
Antibodies
GLG1 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 1.25 - 2.5 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GLG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Gibbon, Bovine, Chicken, Dog, Xenopus, Gorilla, Hamster, Horse, Human, Monkey, Mouse, Rabbit, Rat, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR