Anti-ACAN Rabbit Polyclonal Antibody

71-213
10080-806EA 758.96 CAD
10080-806
Anti-ACAN Rabbit Polyclonal Antibody
ACAN / Aggrecan Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 3 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ACAN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR