Anti-GFPT1 Rabbit Polyclonal Antibody

Supplier: Prosci

71-224
10077-620EA 1042.19 CAD
10077-620
Anti-GFPT1 Rabbit Polyclonal Antibody
Antibodies
GFPT1 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 2.5 - 5 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GFPT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Gibbon, Bovine, Chimpanzee, Chicken, Dog, Guinea pig, Gorilla, Horse, Human, Monkey, Mouse, Orangutan, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR