Anti-PAK7 Rabbit Polyclonal Antibody

Supplier: Prosci

71-234
10080-832EA 983.75 CAD
10080-832
Anti-PAK7 Rabbit Polyclonal Antibody
Antibodies
PAK7 Antibody can be used for ELISAstarting at , and Immunohistochemistry in paraffin embeded tissues starting at 5 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PAK7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human, Orangutan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR