Anti-PTH2R Rabbit Polyclonal Antibody

71-239
10080-842EA 642.26 CAD
10080-842
Anti-PTH2R Rabbit Polyclonal Antibody
PTH2R / PTHR2 Antibody can be used for ELISAstarting at , and Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PTH2R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Bovine, Gorilla, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR