Anti-MAPT Rabbit Polyclonal Antibody

Supplier: Prosci

71-256
10080-870EA 1042.19 CAD
10080-870
Anti-MAPT Rabbit Polyclonal Antibody
Antibodies
MAPT / TAU Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 15 ?g/mL, and Western Blotting starting at 1:500 - 1:1000.

PBS (without Mg2+, Ca2+), pH 7.4, 150 mM NaCl, 0.02% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MAPT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Gibbon, Bovine, Chimpanzee, Dog, Gorilla, Goat, Horse, Human, Monkey, Mouse, Orangutan, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR