Anti-TFRC Rabbit Polyclonal Antibody

Supplier: Prosci

71-277
10075-270EA 1041.86 CAD
10075-270
Anti-TFRC Rabbit Polyclonal Antibody
Antibodies
  • TFRC / Transferrin Receptor Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 μg/mL, and Western Blotting starting at 1:500 - 1:3000.

Protein A Column

PBS, pH 7, 40% Glycerol, 0.01% Thimerosal

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TFRC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR