Anti-FFAR1 Rabbit Polyclonal Antibody

Supplier: Prosci

71-310
10075-330EA 1078.68 CAD
10075-330
Anti-FFAR1 Rabbit Polyclonal Antibody
Antibodies
FFAR1 / GPR40 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL, and Western Blotting starting at 1:500 - 1:3000.

PBS, pH 7, 1% BSA, 20% Glycerol, 0.01% Thimerosal

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: FFAR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR