Anti-CK18 Rabbit Polyclonal Antibody

71-327
10080-930EA 835.8 CAD
10080-930
Anti-CK18 Rabbit Polyclonal Antibody
KRT18 / Cytokeratin 18 Antibody can be used for Immunohistochemistry in frozen tissues starting at 1:100 - 1:200, and Immunohistochemistry in paraffin embeded tissues starting at 1:25 - 1:100.

20 mM Tris-HCl, pH 8.0, 20 mg/ml BSA, 0.05% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: KRT18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR