Anti-MAP3K7 Rabbit Polyclonal Antibody

Supplier: Prosci
71-342
10080-960EA 983.75 CAD
10080-960
Anti-MAP3K7 Rabbit Polyclonal Antibody
Antibodies
MAP3K7 / TAK1 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL, and Western Blotting.

PBS, pH 7.2, 50% glycerol, 0.09% azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MAP3K7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Dog, Xenopus, Guinea pig, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR