To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:mitogen-activated protein kinase kinase kinase 3
- Clonality:Polyclonal
- Gene ID:Q99759
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Antigen symbol:MAP3K3
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:0.05 mL
- Western blot:Yes
- Antigen synonyms:MAP3K3
- Storage temperature:Antibody may be stored short term at 4 °C, long term aliquot and store at –20 °C, avoid freeze thaw cycles.
- Shipping temperature:4 °C
- Immunogen:A 17 residue synthetic peptide based on the mouse MekK3 (residues 2-18) with the cysteine (C) residue added and the peptide coupled to KLH.
- Purification:Antiserum
- Cat. no.:10080-962
Specifications
About this item
MAP3K3 / MEKK3 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 1:100, and Western Blotting starting at 1:5000.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: MAP3K3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat