Anti-TCF7L2 Rabbit Polyclonal Antibody

Supplier: Prosci

71-352
10080-980EA 1041.86 CAD
10080-980
Anti-TCF7L2 Rabbit Polyclonal Antibody
Antibodies
TCF7L2 Antibody can be used for Immunocytochemistry starting at 1 ?g/mL, Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL, Immunoprecipitation and Western Blotting.

PBS, 15 mM sodium azide, approx., pH 7.4.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TCF7L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR