Anti-HMOX1 Rabbit Polyclonal Antibody

Supplier: Prosci

71-357
10075-332EA 983.75 CAD
10075-332
Anti-HMOX1 Rabbit Polyclonal Antibody
Antibodies
HMOX1 / Heme Oxygenase 1 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL, and Western Blotting starting at 1:1000.

PBS, 0.09% azide, 50% glycerol

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HMOX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR