Anti-HSP90AB1 Rabbit Polyclonal Antibody

Supplier: Prosci
71-359
10075-336EA 983.75 CAD
10075-336
Anti-HSP90AB1 Rabbit Polyclonal Antibody
Antibodies
CREB1 / CREB Antibody can be used for Immunohistochemistry starting at 1:50 - 1:100, and Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL,

PBS, pH 7.2, 0.09% azide, 50% glycerol

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HSP90B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR