Anti-HSPA8 Rabbit Polyclonal Antibody

Supplier: Prosci

71-361
10080-988EA 983.75 CAD
10080-988
Anti-HSPA8 Rabbit Polyclonal Antibody
Antibodies
HSP70/HSP73/HSC70/HSPA4/HSPA8 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL, and Western Blotting starting at 1:1000.

PBS, pH 7.2, 0.09% azide, 50% glycerol

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Hspa8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Dog, Drosophila, Fish, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR