Anti-YWHAE Rabbit Polyclonal Antibody

Supplier: Prosci
71-370
10077-646EA 1093.01 CAD
10077-646
Anti-YWHAE Rabbit Polyclonal Antibody
Antibodies
YWHAE / 14-3-3 Epsilon Antibody can be used for ELISA, Immunohistochemistry in paraffin embeded tissues starting at 1:400, and Western Blotting starting at 1:3000.

Serum, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: YWHAE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Chicken, Human, Mouse, Rabbit, Rat, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR