Anti-RPS6KA3 Rabbit Polyclonal Antibody

Supplier: Prosci

71-371
10077-648EA 983.75 CAD
10077-648
Anti-RPS6KA3 Rabbit Polyclonal Antibody
Antibodies
RPS6KA3 / RSK2 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 20 ?g/mL, Immunoprecipitation and Western Blotting starting at 1?g/mL.

PBS, pH 7.2, 50% glycerol, 0.09% azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: RPS6KA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Gibbon, Bovine, Chimpanzee, Chicken, Dog, Gorilla, Hamster, Horse, Human, Monkey, Mouse, Orangutan, Pig, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR