Anti-TXN Rabbit Polyclonal Antibody

Supplier: Prosci
71-382
10074-190EA 1147.67 CAD
10074-190
Anti-TXN Rabbit Polyclonal Antibody
Antibodies
TXN / Thioredoxin / TRX Antibody can be used for ELISA, Immunohistochemistry in paraffin embeded tissues starting at 1:100, and Western Blotting starting at 1:3000.

10 mM sodium HEPES, pH 7.5, 150 mM NaCl, 100 ug/ml BSA, 50% glycerol, 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TXN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR