Anti-PHB Rabbit Polyclonal Antibody

Supplier: Prosci

71-383
10074-192EA 1042.19 CAD
10074-192
Anti-PHB Rabbit Polyclonal Antibody
Antibodies
PHB / Prohibitin Antibody can be used for Immunofluorescence, Immunohistochemistry in paraffin embeded tissues starting at 5 ?g/mL, Immunoprecipitationstarting at 10 - 20?g/mL, and Western Blotting starting at 0.5 - 4?g/mL.

PBS, pH 7.2, 30% glycerol, 0.5% BSA, 0.01% Thimerosal

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PHB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR