Anti-ABCA1 Rabbit Polyclonal Antibody

Supplier: Prosci
71-391
10080-998EA 983.75 CAD
10080-998
Anti-ABCA1 Rabbit Polyclonal Antibody
Antibodies
ABCA1 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 1:100, and Western Blotting starting at 1:500 - 1:5000.

PBS, pH 7.4, 0.02% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ABCA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR