Anti-JAKMIP2 Rabbit Polyclonal Antibody

Supplier: Prosci

7167
10061-788EA 716.01 CAD
10061-788
Anti-JAKMIP2 Rabbit Polyclonal Antibody
Antibodies
JAKMIP2 Antibody: JAKMIP2, also known as neuroendocrine long coiled-coil protein 1 (NECC1), is a member of the JAKMIP family of proteins whose members are thought play a role in JAK1 signaling and the regulation of microtubule cytoskeleton rearrangements. JAKMIP2 is expressed predominantly in neuronal cells but unlike JAKMIP1, JAKMIP2 localizes to the Golgi complex. It has been suggested that JAKMIP2 may play important roles in the control of the regulated secretory pathway.

JAKMIP2 antibody can be used for detection of JAKMIP2 by Western blot at 1 - 2 ?g/mL.

JAKMIP2 Antibody is affinity chromatography purified via peptide column.

JAKMIP2 Antibody is supplied in PBS containing 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: JAKMIP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR