Anti-BCL3 Rabbit Polyclonal Antibody

Supplier: Prosci
7183
10062-310EA 716.01 CAD
10062-310
Anti-BCL3 Rabbit Polyclonal Antibody
Antibodies
BCL3 Antibody: The proto-oncogene Bcl-3, believed to be involved in certain human B cell leukemias, encodes a protein that contributes to the regulation of transcriptional activation of NF-κ-B target genes. BCL3 contains seven ankyrin repeats, which are most closely related to those found in I-κ-B proteins. The expression of this gene can be induced by NF-κ-B, which forms a part of the autoregulatory loop that controls the nuclear residence of p50 NF-κ-B. It contributes to the regulation of cell proliferation. A chromosomal aberration involving BCL3 may be a cause of B-cell chronic lymphocytic leukemia.

BCL3 antibody can be used for detection of BCL3 by Western blot at 1 - 2 ?g/mL.

BCL3 Antibody is affinity chromatography purified via peptide column.

BCL3 Antibody is supplied in PBS containing 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: BCL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR