Anti-FOXP3 Rabbit Polyclonal Antibody

Supplier: Prosci
7187
10062-314EA 716.01 CAD
10062-314
Anti-FOXP3 Rabbit Polyclonal Antibody
Antibodies
FOXP3 Antibody: FOXP3 is a member of the forkhead/winged-helix family of transcriptional regulators. FOXP3 acts as a repressor of transcription and regulates T cell activation, with its overexpression in CD4 T cells leading to an attenuation of activation-induced cytokine production and proliferation. In regulatory T (Treg) cells, FOXP3 is essential for Treg suppressor function and its expression leads to the repression of IL-17 expression. Genetic mutations involving FOXP3 are the cause of immunodeficiency polyendocrinopathy, enteropathy, X-linked syndrome (IPEX), also known as X-linked autoimmunity-immunodeficiency syndrome.

FOXP3 antibody can be used for detection of FOXP3 by Western blot at 1 - 2 ?g/mL.

FOXP3 Antibody is affinity chromatography purified via peptide column.

FOXP3 Antibody is supplied in PBS containing 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: FOXP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR