Anti-STAT1 Rabbit Polyclonal Antibody

7195
10061-802EA 630.03 CAD
10061-802
Anti-STAT1 Rabbit Polyclonal Antibody
STAT1 Antibody: STATs (signal transducers and activators of transcription) are a family of cytoplasmic latent transcription factors that are activated to regulate gene expression in response to a large number of extracellular signaling polypeptides including cytokines, interferons, and growth factors. After phosphorylation by JAK tyrosine kinases, STATs enter the nucleus to regulate transcription of many different genes. Among the seven STATs (STAT1, STAT2, STAT3, STAT4, STAT5a, STAT5b, and STAT6), STAT1, STAT3, STAT5a, and STAT5b have a wide activation profile. STAT1 is activated by many different ligands including IFN family (IFN-beta, IFN-beta, IFN-gamma and IL-10), gp130 family (IL-6, IL-11, LIF, CNTF, and G-CSF), and receptor tyrosine kinases (EGF, PDGF, and CSF-1).

STAT1 antibody can be used for detection of STAT1 by Western blot at 1 - 2 ?g/mL.

STAT1 Antibody is affinity chromatography purified via peptide column.

STAT1 Antibody is supplied in PBS containing 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: STAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR