Anti-PRKCDBP Rabbit Polyclonal Antibody

Supplier: Prosci

7269
10061-842EA 716.01 CAD
10061-842
Anti-PRKCDBP Rabbit Polyclonal Antibody
Antibodies
PRKCDBP Antibody: The protein kinase C delta (PKC-δ) binding protein (PRKCDBP), also known as cavin-3, is a member of the cavin family of proteins that are involved in caveolin formation and regulation. PRKCDBP was initially identified in a screen of cultured cell lines for proteins that were strongly induced by serum starvation. Studies indicate that PRKCDBP binds not only to PKC-δ but also to caveolin-1 and helps regulate caveolin traffic and function. Similar to other members of the cavin family, the expression of PRKCDBP was found to be down-regulated in various cancer cell lines, suggesting a possible tumor suppressor function of PRKCDBP.

PRKCDBP antibody can be used for detection of PRKCDBP by Western blot at 1 - 2 ?g/mL.

PRKCDBP Antibody is affinity chromatography purified via peptide column.

PRKCDBP Antibody is supplied in PBS containing 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PRKCDBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR