Anti-MTERF3 Rabbit Polyclonal Antibody

Supplier: Prosci
7281
10062-538EA 716.01 CAD
10062-538
Anti-MTERF3 Rabbit Polyclonal Antibody
Antibodies
MTERFD1 Antibody: Members of the mTERF (mitochondrial transcription termination factor) family, are mitochondrial proteins that are believed to be transcription termination factors. MTERFD1 is composed of three leucine zippers that form a three-stranded coiled-coil that binds to DNA. It binds promoter DNA and acts as a mitochondrial transcription regulator. It has been suggested that only the phosphorylated form of MTERFD1 has transcription termination activity. A closely related mTERF family member, MTERFD3, is believed to be involved in cell cycle regulation and cell growth by modulating mitochondrial transcription.

MTERFD1 Antibody can be used for detection of MTERFD1 by Western blot starting at 1 ?g/mL. Antibody can also be used for Immunocytochemistry starting at 5 ?g/mL. For immunofluorescence start at 20 ?g/mL.

MTERFD1 Antibody is affinity chromatography purified via peptide column.

MTERFD1 Antibody is supplied in PBS containing 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MTERFD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR