Anti-MTERF4 Rabbit Polyclonal Antibody

Supplier: Prosci
7283
10061-854EA 716.01 CAD
10061-854
Anti-MTERF4 Rabbit Polyclonal Antibody
Antibodies
MTERFD2 Antibody: Members of the mTERF (mitochondrial transcription termination factor) family, are mitochondrial proteins that are believed to be transcription termination factors. MTERFD2 is targeted to the mitochondria and is ubiquitously expressed, with highest expression levels in fore- and midbrain, diencephalon, spinal cord, tongue, lung liver and kidney. MTERFD2 has been suggested to play a role in organ differentiation during embryogenesis. A closely related mTERF family member, MTERFD3, is believed to be involved in cell cycle regulation and cell growth by modulating mitochondrial transcription.

MTERFD2 Antibody can be used for detection of MTERFD2 by Western blot at 1 ?g/mL.

MTERFD2 Antibody is affinity chromatography purified via peptide column.

MTERFD2 Antibody is supplied in PBS containing 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MTERFD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR