Anti-C1orf64 Rabbit Polyclonal Antibody

Supplier: Prosci

7289
10061-860EA 715.82 CAD
10061-860
Anti-C1orf64 Rabbit Polyclonal Antibody
Antibodies
ERRF Antibody: The estrogen receptor-related factor (ERRF) is a recently identified protein whose expression is upregulated in multiple cell lines and primary breast cancer tumors. Increased ERRF expression was significantly associated with ER or progesterone receptor (PR) positivity even in HER2-negative tumors. This higher ERRF expression correlated with better disease-free survival and overall survival. In two ER-positive breast cancer cell lines, knockdown of ERRF suppressed cell growth in vitro and tumorigenesis in xenograft models, suggesting that ERRF plays a role in estrogen-ER-mediated breast cancer cell growth and may thus be a potential therapeutic target.

ERRF Antibody can be used for detection of ERRF by Western blot at 1 ?g/mL.

ERRF Antibody is affinity chromatography purified via peptide column.

ERRF Antibody is supplied in PBS containing 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: C1orf64
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR