Anti-TLX2 Rabbit Polyclonal Antibody

Supplier: Prosci

7373
10062-540EA 716.01 CAD
10062-540
Anti-TLX2 Rabbit Polyclonal Antibody
Antibodies
TLX2 Antibody: The Hox proteins play a role in patterns of embryonic development and cellular differentiation by regulating downstream target genes. TLX2, also known as homeobox11-like 1 (HOX11L1), is a DNA-binding nuclear transcription factor that appears to be crucial for the development of the peripheral nervous system. TLX2 is a downstream target of regulation by both PHOX2A and PHOX2B proteins during intestinal neuronal differentiation. In humans, loss-of-function of TLX2 may play a role in tumorigenesis of gastrointestinal stromal tumors.

TLX2 antibody can be used for detection of TLX2 by Western blot at 0.5 - 1 ?g/mL.

TLX2 Antibody is affinity chromatography purified via peptide column.

TLX2 Antibody is supplied in PBS containing 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TLX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR