Anti-N Rabbit Polyclonal Antibody

Supplier: Prosci
7417
10061-924EA 716.01 CAD
10061-924
Anti-N Rabbit Polyclonal Antibody
Antibodies
La Crosse Virus Nucleocapsid Antibody: La Crosse virus (LCV) is a single-stranded RNA virus that can cause encephalitis in humans and is transmitted predominantly by mosquitoes. It is often mistaken for herpes simplex encephalitis. During human infections, LCV initially presents as a nonspecific summertime illness with fever, headache, nausea, vomiting and lethargy. Death occurs in less than 1% of clinical cases.

La Crosse virus nucleocapsid antibody can detect 10 ng La Crosse virus nucleocapsid peptide in ELISA at 1 ?g/mL.

La Crosse Virus Nucleocapsid Antibody is affinity chromatography purified via peptide column.

La Crosse virus nucleocapsid Antibody is supplied in PBS containing 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: N
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR