Anti-KLK4 Rabbit Polyclonal Antibody

Supplier: Prosci
7445
10061-946EA 716.01 CAD
10061-946
Anti-KLK4 Rabbit Polyclonal Antibody
Antibodies
Kallikreins (KLKs) belong to the serine protease family of proteolytic enzymes having diverse physiological functions. Kallikrein 4 (KLK4), also known as prostase or EMSP1 (enamel matrix serine protease 1), contains one peptidase S1 domain and the expression in prostate is regulated by hormone (1,2). Many kallikreins are implicated in carcinogenesis and have potential as novel cancer and other disease biomarkers. KLK4 may have additional roles such as functioning as one of the two major enamel proteases identified that process enamel matrix proteins. Defects in KLK4 are the cause of amelogenesis imperfecta hypomaturation type 2A1 (AI2A1).

KLK4 antibody can be used for detection of KLK4 by Western blot at 1 - 2 µg/ml.

KLK4 Antibody is affinity chromatography purified via peptide column.

KLK4 Antibody is supplied in PBS containing 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: KLK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR