Anti-DDC Rabbit Polyclonal Antibody

Supplier: Prosci
7447
10061-948EA 716.01 CAD
10061-948
Anti-DDC Rabbit Polyclonal Antibody
Antibodies
DOPA decarboxylase (DDC) belongs to the group II decarboxylase family of proteins. It is an important protein in the catecholamine biosynthesis pathway. DDC catalyzes the second reaction in the biosynthesis of catecholamines, trace amines and serotonin (1,2). It can form a homodimer and is expressed in the central nervous system. DDC can be used as markers for dopaminergic, noradrenergic and serotonergic neurons in a variety of applications including depression, schizophrenia, Parkinson's disease, neuroendocrine tumors and drug abuse. Defects in DDC gene may cause the autosomal recessive disorder AADC deficiency.

DDC antibody can be used for detection of DDC by Western blot at 1 - 2 µg/ml.

DDC Antibody is affinity chromatography purified via peptide column.

DDC Antibody is supplied in PBS containing 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DDC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR