Anti-CEACAM5 Rabbit Polyclonal Antibody

Supplier: Prosci
7453
10061-954EA 716.01 CAD
10061-954
Anti-CEACAM5 Rabbit Polyclonal Antibody
Antibodies
CEACAM5 is a member of the carcinoembryonic antigen (CEA) family, which belongs to the immunoglobulin superfamily (1,2). CEACAM proteins are involved in cell-cell recognition and modulate cellular processes that range from the shaping of tissue architecture and neovascularization to the regulation of insulin homeostasis and T-cell proliferation. CEACAM5 has been shown to be overexpressed in a majority of carcinomas and has been suggested to be a intercellular adhesion molecule involved in cancer invasion and metastasis.

CEACAM5 antibody can be used for detection of CEACAM5 by Western blot at 1 - 2 µg/ml.

CEACAM5 Antibody is affinity chromatography purified via peptide column.

CEACAM5 Antibody is supplied in PBS containing 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CEACAM5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR