Anti-IL36G Rabbit Polyclonal Antibody

Supplier: Prosci
7509
10061-998EA 716.01 CAD
10061-998
Anti-IL36G Rabbit Polyclonal Antibody
Antibodies
IL-36G is is a member of the interleukin 1 cytokine family whose gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. IL-36G is thought to activate the NF-kappaB pathway through IL-1 receptor family members IL-1RL2 and IL-1RAcP. Like the related proteins IL-36A and IL-36B, IL-36G requires post-translational processing for full agonist activity, but the cleavage mechanism is currently unknown. IL-36G expression is stimulated by interferon-gamma, tumor necrosis factor-alpha and interleukin 1. The IL-36 cytokines have been suggested to amplify Th1 responses by enhancing proliferation and Th1 polarization of naive CD4+ T cells.

IL-36G antibody can be used for detection of IL-36G by Western blot at 1 - 2 ?g/ml.

IL-36G antibody is affinity chromatography purified via peptide column.

IL-36G antibody is supplied in PBS containing 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: IL36G
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR