Anti-ENHO Rabbit Polyclonal Antibody

Supplier: Prosci
7527
10062-006EA 716.01 CAD
10062-006
Anti-ENHO Rabbit Polyclonal Antibody
Antibodies
ADROPIN is a recently identified protein that has been implicated in the maintenance of energy homeostasis and insulin resistance. ADROPIN expression is regulated by energy status and dietary nutrient content and is altered by obesity and regulates the expression of hepatic lipogenic genes and adipose tissue peroxisome proliferator-activated receptor gamma (PPAR-gamma). ADROPIN levels increase with dietary fat content. ADROPIN has also been proposed to play a role in the regulation of endothelial function.

ADROPIN antibody can be used for detection of ADROPIN by Western blot at 1 - 2 ?g/ml. Antibody can also be used for Immunohistochemistry starting at 5 ?g/mL. For immunofluorescence start at 20 ?g/mL.

ADROPIN antibody is affinity chromatography purified via peptide column.

ADROPIN antibody is supplied in PBS containing 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ENHO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR