Anti-KDM1B Rabbit Polyclonal Antibody

Supplier: Prosci
7535
10062-010EA 716.01 CAD
10062-010
Anti-KDM1B Rabbit Polyclonal Antibody
Antibodies
Flavin-dependent histone demethylases regulate histone lysine methylation, an epigenetic mark that regulates gene expression and chromatin function . KDM1B is a recently identified histone H3K4 demethylase that is required to establish maternal genomic imprints; targeted disruption of the KDM1B gene in mice led to lethality in their embryos, suggesting demethylation of H3K4 is critical for establishing the DNA methylation imprints. KDM1B has also been shown to play a role in the transcription elongation in several genes.

KDM1B antibody can be used for detection of KDM1B by Western blot at 1 - 2 ?g/ml.

KDM1B antibody is affinity chromatography purified via peptide column.

KDM1B antibody is supplied in PBS containing 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: KDM1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR