Anti-AMIGO1 Rabbit Polyclonal Antibody

Supplier: Prosci
7565
10062-016EA 716.01 CAD
10062-016
Anti-AMIGO1 Rabbit Polyclonal Antibody
Antibodies
The amphoterin-induced gene and ORF (AMIGO1) protein is a brain-enriched, glycosylated transmembrane immunoglobulin (Ig) superfamily protein with six extracellular leucine-rich repeats (LRRs) and one Ig-like domain. It and the related proteins AMIGO2 and AMIGO3 are thought to be cell adhesion molecules expressed on fiber tracts of neuronal tissues and participate in their formation. AMIGO1 has also been suggested to play important roles in dendritic outgrowth during development and could modulate the survival of developing and adult neurons.

AMIGO1 antibody can be used for detection of AMIGO1 by Western blot at 1 - 2 ?g/ml. Antibody can also be used for Immunohistochemistry starting at 5 ?g/mL. For immunofluorescence start at 20 ?g/mL.

AMIGO1 antibody is affinity chromatography purified via peptide column.

AMIGO1 antibody is supplied in PBS containing 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: AMIGO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR