The ULBPs are unusual members of the extended MHC class I superfamily because they do not contain the alpha 3 domain and they lack a transmembrane domain. They are known to bind to human NKG2D and activate multiple signaling pathways such as JAK2, STAT5, ERK and PI3K/Akt signal transduction pathways in NK / T cells, resulting in the production of cytokines and chemokines. ULBP1 is expressed on some tumor cells and has been implicated in tumor surveillance.
ULBP1 antibody can be used for detection of ULBP1 by Western blot at 1 - 2 ?g/ml.
ULBP1 antibody is affinity chromatography purified via peptide column.
ULBP1 antibody is supplied in PBS containing 0.02% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ULBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse