Anti-ARR3 Rabbit Polyclonal Antibody

Supplier: Prosci
7643
10062-048EA 715.82 CAD
10062-048
Anti-ARR3 Rabbit Polyclonal Antibody
Antibodies
Arrestin 3 (ARR3) belongs to the Arrestin family of proteins that function as negative regulators of G protein-coupled receptor (GPCR) signaling (1,2). ARR3 binds the c-Jun N-terminal kinase 3 (JNK3) and scaffolds the apoptosis signal-regulating kinase 1 (ASK1)-MAPK kinase4-JNK3 cascade in a receptor-independent fashion, promoting JNK3 phosphorylation. ARR3 also mediates the internalization of the C-C chemokine receptor CCR7 upon the binding of its ligand CCL19, but not CCL21, suggesting that it helps mediate CCR7 receptor desensitization.

ARR3 antibody can be used for detection of ARR3 by Western blot at 0.5 - 1 ?g/ml.

ARR3 antibody is affinity chromatography purified via peptide column.

ARR3 antibody is supplied in PBS containing 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ARR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR